Learn More
CELLTREAT Scientific Products 5mL Aspirating Pipet, Individual Paper/Plastic Wrapper Packed in Bags, Sterile pipette
SDP
Supplier:  CELLTREAT Scientific Products 229265
Encompass_Preferred
CELLTREAT Scientific Products 5mL Aspirating Pipet. Ideal for aspiration of media and sample waste removal. Supplied without graduations or plugs. Rnase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 50 individually wrapped/bag, 200/case.
Catalog No. 50-202-131