Learn More
Thomas Scientific 5 mL Posi-Click Microcentrifuge Tubes, Assorted Colors, 200/pk
SDP
Supplier:  Thomas Scientific 1149Y06
Encompass
5mL Posi-Click Graduated Microcentrifuge tubes feature large, easy-to-open/close cap with the assurance of an audible "click" to know that your sample is secure. Fits all standard 5.0mL centrifuge rotors, racks, and storage boxes.
Catalog No. NC1904825