Learn More
Chemglass Life Sciences Chemglass 5 mL Centrifuge Tube, Screw Cap, Non-Sterile, 500/Pack (CLS-4298-05)
SDP
Supplier:  Chemglass Life Sciences CLS429805
Encompass_Preferred
Chemglass 5 mL centrifuge tubes are clear, low-binding polypropylene macrotubes supplied bulk-packed with separate screw caps. They provide reliable sealing with a single 360° turn, dual graduations for accurate dispensing, and compatibility with standard centrifuge rotors and laboratory equipment. Useful from -86°C to 110°C and rated to withstand up to 25,000 x g, these non-sterile tubes are suitable for routine sample processing and storage.
  • 5 mL capacity.
  • Virgin polypropylene with maximum clarity and low binding.
  • Non-sterile, bulk packed with caps supplied separately.
  • 500 tubes per pack.
  • 16 mm tube diameter, 20 mm cap outer diameter, 56 mm total length.
  • Dual graduations at 0.2 mL and 0.5 mL intervals.
  • Withstands up to 25,000 x g.
  • Usable temperature range -86°C to 110°C.
  • RNase- and DNase-free.
  • Compatible with standard centrifuges, racks, and thermal shakers.
Catalog No. NC1993582