Learn More
Grainger Graduated Cylinders
Supplier:  Grainger 55EZ50
Encompass_Preferred
The website features a variety of graduated cylinders made of glass or plastic for sale. These laboratory instruments are used to accurately measure the volume of a liquid. They have graduated markings along the length of the cylinder to indicate the volume measurement. The cylinders come in various sizes, with capacities ranging from 10mL to 2000mL. They have a narrow, cylindrical shape with a pouring lip. The cylinders are offered by various brands like Pyrex, Kimble, and Wheaton. They can have subdivisions as fine as 0.1mL for precise measurements. The graduated cylinders are an essential tool for chemistry, biology, and other lab work.
Catalog No. NC2088210