Learn More
Bellco Glass, Inc. 5mL Macro Centrifuge Tube
SDP
Supplier:  Bellco Glass, Inc. 229449000
Encompass_Preferred
Description5mL Macro Centrifuge Tubes offer the convenience of a micro centrifuge tube with the ability to process sample volumes up to 5mLTube diameter and shape is the same as a standard 15mL conical bottom centrifuge tube, enabling use with existing equipmentTubes have frosted marking spot and engraved graduationsAttached flat top cap is easy to open and close with one handCenter of the cap includes a thin “membrane” area designed for easy puncture and access by a syringe/needle Pack Description20/Re-sealable BagCase Quantity100SterileYesNon-pyrogenicYesAutoclavableYesPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume5mLGraduatedYesGraduation Increments (Subdivisions)0.2mLMax Force (RCF)g25000RNase/DNase FreeYes
Catalog No. 50-233-0267