Learn More
MSC ANSELL NITRILE DISP GLOVE
Supplier:  MSC 89305445
Encompass
Series TouchNTuff Disposable Gloves: Size X-Large, 4.9 mil, Uncoated-Coated Nitrile, Industrial Grade, Unpowdered
Catalog No. 50-273-6555