Learn More
Sarstedt Inc 5ML PP TUBE 75X13MM
Supplier:  Sarstedt Inc 60.504.021
Encompass_Preferred
Screw cap tube, 5 ml, 75 x 13 mm, round base, PP, without closure, 1,000 pcs./bag
Catalog No. NC1053127
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product