Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences 5 mL Transport Tubes With HDPE Caps, Sterile, 50/Pack
SDP

Supplier:  Chemglass Life Sciences CGN474205

Encompass_Preferred

Sterile 5 mL transport tubes designed for secure sample storage, freezing, and transport in laboratory and clinical settings. Manufactured for low bioburden and leak-resistant sealing suitable for air shipment.

  • Capacity 5 mL.
  • USP VI polypropylene tube material.
  • HDPE caps provide a secure seal.
  • Electron beam sterilized (SAL = 10⁻⁶).
  • Produced in a 100,000-level clean workshop.
  • DNAse-free, RNAse-free, pyrogen-free, endotoxin-free.
  • Self-standing design for stability.
  • Seal maintains integrity at negative pressure 95 kPa.
  • Packaged 50 per pack, 1,000 per case.

Catalog No. 50-273-3405


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.