Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences Chemglass 5 mL Sterile Transport Tubes with Pink Capture Caps, 50/pack (CGN-4742-05P)
SDP

Supplier:  Chemglass Life Sciences CGN474205P

Encompass_Preferred

Sterile 5 mL transport tubes with pink capture caps, made from USP VI polypropylene with HDPE caps. Designed for sample storage, transport, and freezing in clinical, environmental, food testing, and research laboratories. Packaged sterile and certified free of DNase, RNase, pyrogens, and endotoxins.

  • 5 mL polypropylene tube volume.
  • Pink capture caps for secure closure.
  • Electron beam sterilized; SAL 10^-6.
  • DNase-, RNase-, pyrogen-, and endotoxin-free.
  • Pack quantities: 50 per pack, 1,000 per case.
  • Self-standing design for stable bench placement.
  • Temperature range approximately -196°C to 121°C.
  • HDPE caps with sealing performance tested to withstand negative pressure of 95 kPa.

Catalog No. 50-273-3404


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.