Learn More
Ace Glass, Inc. 5MM JOINTS (GB)
SDP
Supplier:  Ace Glass, Inc. 764602
Encompass_Preferred
5mm glass o-ring seal joint tooled with a groove to take a -110 FETFE o-ring, and to provide a greaseless, vacuum-tight connection. Supplied with one o-ring, which may be used up to 230°C.
Catalog No. 50-975-234