Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5MPN | 47208-82-2 | 99.9% | 305.33 g/mol | C15H19N3O4 | 50MG
SDP

Supplier:  Medchemexpress LLC HY12398150MG

Encompass_Preferred

5MPN is a selective small-molecule inhibitor of 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 4 (PFKFB4) used in biochemical and cellular research to probe tumor glucose metabolism and cell proliferation. It is supplied as a high-purity solid suitable for laboratory research applications.

  • Selective inhibitor of PFKFB4 for metabolic studies.
  • Potent activity in cell proliferation assays.
  • High purity (99.89%).
  • Molecular weight 305.33 g/mol; formula C15H19N3O4.
  • Store at -20 °C; supplied as a 50 mg pack.

Catalog No. 50-002-38901


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.