Learn More
Cayman Chemical 5PhosphoDrIbose 1dIphosph 10mg
SDP
Supplier:  Cayman Chemical 1889710 MG
Encompass_Preferred
A natural intermediate involved in the pentose phosphate pathway leading to purine, pyrimidine, and histidine metabolism
Catalog No. 50-298-4442