Learn More
USA Scientific Inc 5-place Upright freezer rack
Supplier:  USA Scientific Inc 26510900
Encompass
5-place (5 long x 1 high) upright freezer rack for multiple well plates, 18 1/2 in L x 5 1/2 in W x 9 1/4 in H, stainless steel
Catalog No. 50-274-5010