Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (5R)-5-hydroxytriptolide | 583028-68-6 | MFCD15147006 | 99.7% | 376.40 g·mol⁻¹ | C20H24O7 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1071011MG

Encompass_Preferred

5R-5-Hydroxytriptolide (CAS 583028-68-6) is a triptolide-derived small molecule used in research for its immunosuppressive properties. It exhibits reduced cell cytotoxicity compared with related compounds and is supplied as a high-purity reagent for in vitro studies.

  • Immunosuppressive activity useful for in vitro assays.
  • Lower cell cytotoxicity compared with related compounds.
  • High purity (99.7%).
  • Molecular formula C20H24O7.
  • Molecular weight 376.40 g·mol⁻¹.
  • Available in small-scale research quantities (1 mg, 5 mg, 10 mg, 50 mg).

Catalog No. 50-002-39774


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.