Learn More
CPC Scientific 5TAMRA-Ala-Met-Arg-Leu-Glu-Arg-Gln-Asp-Ser-Ile-Phe-Tyr-Pro-Lys-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PROK050B
Encompass
ONE-LETTER SEQUENCE: 5TAMRA-AMRLERQDSIFYPK-NH2MOLECULAR FORMULA: C103H145N25O25S1MOLECULAR WEIGHT:2165.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0314