Learn More
CPC Scientific 5TAMRA-Glu-Phe-Pro-Ile-Tyr-Asp-Phe-Leu-Pro-Ala-Lys-Lys-Lys-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PROK052B
Encompass
ONE-LETTER SEQUENCE: 5TAMRA-EFPIYDFLPAKKK-NH2MOLECULAR FORMULA: C104H139N19O22MOLECULAR WEIGHT:2007.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0318