Learn More
Bevco Precision Manufacturing Co Doral Mid Height ESD/ISO 5 Cleanroom Blue Vinyl Chair; Non-Tilt; Adjustable Footring; Polished Aluminum Base w/Welded Footring
Supplier:  Bevco Precision Manufacturing Co 5350E3BLV
Encompass_Preferred
BEVCO’s Doral ESD/ISO 5 Cleanroom Series models are designed to meet specific particulate goals of ISO 14644-1 and the ESD requirements of ANSI/ESD STM12.1-2019 delivering a combination ESD/Cleanroom seating solution for highly technical environments. Specifically, users that have a petite-to-average build will truly enjoy this seating experience.This non-tilt model features manual back and seat height adjustments for increased comfort during extended sitting durations.Fully adjustable production-style back has a durable scuff resistant back pan to protect the upholstery.A waterfall seat promotes lower body circulation, improving overall wellness.The pneumatic height adjustment allows you to easily change your seat height from 19" - 26.5".Upholstered ESD vinyl meets CAL 117 standards.The chair features a sturdy 27" diameter polished aluminum base with welded footring.Model comes standard with ESD mushroom glides. Optional ESD casters can be ordered separately.
Catalog No. NC2125906