Learn More
Medchemexpress LLC 5Z-[(2E)-2-methyl-3-phenyl-d5-2-propen-1-ylidene]-4-oxo-2-thioxo-3-thiazolidineacetic acid | 2755644-33-6 | >99.0% | 324.4 g·mol⁻1 | C15H8D5NO3S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW6539445MG
Encompass_Preferred
Epalrestat-d5 is a deuterium-labeled (d5) analogue of epalrestat supplied as a solid reference standard for analytical use. It is intended as an internal standard for quantitative GC- or LC-MS assays, providing high isotopic purity and consistent mass for accurate measurements.
  • Designed as an internal standard for GC- or LC-MS quantification.
  • Deuterium-labeled (d5) isotopologue for clear mass separation from the parent compound.
  • High deuteration purity to minimize isotopic interference in assays.
  • Slightly soluble in DMSO and methanol for use in common assay solvents.
  • Well characterized molecular formula C15H8D5NO3S2 and formula weight 324.4 g·mol⁻1.
Catalog No. 50-002-84443