Learn More
Medchemexpress LLC 5Z-7-oxozeaenol | 253863-19-3 | MFCD12912700 | 99.6% | 362.37 g/mol | C19H22O7 | 5 MG
Supplier: Medchemexpress LLC HY126865MG
5Z-7-Oxozeaenol is a natural, fungal-derived small molecule that acts as a potent, irreversible, and selective inhibitor of TAK1 and VEGF-R2. It is used in biochemical and cellular research to probe TAK1- and VEGFR2-mediated signaling; reported IC50 values are 8 nM for TAK1 and 52 nM for VEGF-R2. The compound is supplied as a solid with high purity suitable for research applications.
- Potent, irreversible inhibition of TAK1 (IC50 = 8 nM).
- Selective activity against VEGF-R2 (IC50 = 52 nM).
- High purity suitable for biochemical and cellular assays.
- Solid form convenient for handling and storage.
- Useful for studying MAPKKK-mediated signaling pathways.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.