Learn More
Cayman Chemical 5Z8Z11ZHeptadecatrIe100ug
SDP
Supplier:  Cayman Chemical 39690100 UG
Encompass_Preferred
An internal standard for the quantification of 5(Z),8(Z),11(Z)-heptadecatrienoic acid by GC- or LC-MS
Catalog No. 50-291-4760