Learn More
Medchemexpress LLC 6-[1-(2-fluoropyridin-3-yl)-5-methyl-1H-1,2,3-triazol-4-yl]quinoline | 864863-72-9 | 99.9% | 305.31 | C17H12FN5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10038250MG
Encompass_Preferred
FPTQ is a potent metabotropic glutamate receptor 1 (mGluR1) antagonist with low-nanomolar inhibitory potency (human IC50 = 6 nM; mouse IC50 = 1.4 nM). It has reported antioxidant and anti-inflammatory activity in vitro and in vivo and is supplied as a solid research compound suitable for pharmacology and neuroscience studies.
  • Low-nanomolar mGluR1 antagonist potency, enabling sensitive receptor modulation studies.
  • Demonstrated antioxidant and anti-inflammatory effects in vitro and in vivo.
  • High purity (99.9%), supporting reproducible experimental results.
  • Available in small solid quantities for compound screening and animal dosing.
  • Soluble in DMSO; protocols available for in vivo formulation.
  • Stable under recommended storage conditions to preserve integrity.
Catalog No. 50-002-13984