Learn More
Sklar Instruments Single Use, Econo-Sterile, Rochester-Pean Forceps, Sklar 6-1/4 in., Blunt, Straight, Horizontally Serrated, Stainless Steel, Disposable. CS of 25.
SDP
Supplier:  Sklar Instruments 962545
Encompass_Preferred
Single Use, Econo-Sterile, Rochester-Pean Forceps,Sklar, 6-1/4 in., Blunt,Straight, Horizontally Serrated, Stainless Steel, Disposable. CS of 25.Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
Catalog No. 50-109-4260