Learn More
Cayman Chemical 6 15dIketo13 14dIhydro Pro 5mg
SDP
Supplier:  Cayman Chemical 152705 MG
Encompass_Preferred
A metabolite of PGI2; enhances intracellular cAMP and cholesterol catabolism in bovine arterial smooth muscle cells
Catalog No. 50-296-9617