Learn More
Medchemexpress LLC 6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidin-4(1H)-one | 1628317-84-9 | 234.20 g/mol | C8H5F3N2OS | 500 MG

Supplier: Medchemexpress LLC HYW071559500MG
6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidin-4(1H)-one is a small-molecule synthetic intermediate used in medicinal chemistry for the preparation of thienopyrimidine-derived active compounds. Intended for research use only.
- Used as a drug intermediate for synthesis of active compounds.
- Intended for research use only.
- Available in multiple pack sizes, including 500 mg.
- Molecular formula: C8H5F3N2OS.
- Molecular weight: 234.20 g/mol.
- CAS number: 1628317-84-9.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.