Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-(2-(2,4-Difluorophenoxy)-5-((ethylsulfonyl)methyl)pyridin-3-yl)-8-methylpyrrolo[1,2-a]pyrazin-1(2H)-one | 2411226-02-1 | 99.7% | 10 MM * 1 ML
SDP

Catalog No. 5000340701
Encompass_Preferred

BET bromodomain inhibitor 1 is an orally active, selective bromodomain and extra-terminal (BET) bromodomain inhibitor with an IC50 of 2.6 nM for BRD4. It binds to BRD2(2), BRD3(2), BRD4(1), BRD4(2), and BRDT(2) with high affinities (Kd values of 1.3 nM, 1.0 nM, 3.0 nM, 1.6 nM, 2.1 nM, respectively). This inhibitor also demonstrates anti-cancer activity.

  • Orally active, selective bromodomain and extra-terminal (BET) bromodomain inhibitor.
  • Binds to BRD2(2), BRD3(2), BRD4(1), BRD4(2), and BRDT(2) with high affinities.
  • Exhibits anti-cancer activity.
  • Demonstrates antiproliferative activity against various human leukemia and myeloma cell lines.
  • Induces G1-phase cell cycle arrest and reduces c-Myc, BCL-2, and CDK6 expressions in MV-4-11 cells.
  • Offers high selectivity for the BET bromodomain family.
  • Shows strong antitumor activities in an MV4-11 mouse xenograft model.

Catalog No. 50-003-40701 Supplier Medchemexpress LLC Supplier No. HY13106110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BET bromodomain inhibitor 1 is an orally active, selective bromodomain and extra-terminal (BET) bromodomain inhibitor with an IC50 of 2.6 nM for BRD4. It binds to BRD2(2), BRD3(2), BRD4(1), BRD4(2), and BRDT(2) with high affinities (Kd values of 1.3 nM, 1.0 nM, 3.0 nM, 1.6 nM, 2.1 nM, respectively). This inhibitor also demonstrates anti-cancer activity.

  • Orally active, selective bromodomain and extra-terminal (BET) bromodomain inhibitor.
  • Binds to BRD2(2), BRD3(2), BRD4(1), BRD4(2), and BRDT(2) with high affinities.
  • Exhibits anti-cancer activity.
  • Demonstrates antiproliferative activity against various human leukemia and myeloma cell lines.
  • Induces G1-phase cell cycle arrest and reduces c-Myc, BCL-2, and CDK6 expressions in MV-4-11 cells.
  • Offers high selectivity for the BET bromodomain family.
  • Shows strong antitumor activities in an MV4-11 mouse xenograft model.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.