Learn More
Medchemexpress LLC 6-[2-[4-[(4-fluorophenyl)-phenylmethylidene]piperidin-1-yl]ethyl]-7-methyl-thiazolo[3,2-a]pyrimidin- | 93076-89-2 | MFCD00055114 | 99.8% | 459.58 | C27H26FN3OS | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10761350MG
Encompass_Preferred
R 59-022 is a small-molecule diacylglycerol kinase (DGK) inhibitor for biochemical and cellular research. It modulates protein kinase C activity, acts as a 5-HT receptor antagonist, and potentiates thrombin-induced diacylglycerol production in platelets. It is supplied as a solid for in vitro studies.
  • Diacylglycerol kinase inhibitor with reported IC50 ≈2.8 μM.
  • Modulates protein kinase C and 5-HT receptor activity.
  • High purity (99.8% by HPLC).
  • Molecular weight 459.58 g/mol.
  • Supplied as small-scale solid suitable for in vitro research.
Catalog No. 50-002-41617