Learn More
Medchemexpress LLC 6-(2-aminophenoxy)-1,2-benzisothiazol-3-amine dihydrochloride | 613263-00-6 | MFCD30179737 | 99.9% | 330.23 g·mol⁻1 | C13H13Cl2N3OS | 25 MG
SDP
Supplier:  Medchemexpress LLC HY103639A25MG
Encompass_Preferred
M62812 is a small-molecule Toll-like receptor 4 (TLR4) inhibitor supplied as a solid research reagent for studies of innate immune signaling and sepsis models. It is provided in small vial sizes and as ready-to-use DMSO solutions for laboratory experiments.
  • Small-molecule TLR4 inhibitor for research use.
  • High purity: 99.9% (reported by supplier).
  • Solid, white to off-white appearance.
  • Available in small vial sizes, including 25 MG.
  • Storage: solid at 4°C; solutions stable at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-44691