Learn More
Medchemexpress LLC 6-(2-hydroxyethoxy)-1-hexanol | 23684-16-4 | 162.23 | 25 MG

Supplier: Medchemexpress LLC 25MGHY22245
6-(2-Hydroxyethoxy)-1-hexanol is a drug intermediate for the synthesis of various active compounds. This product is suitable for laboratory research and development.
- Target: Drug intermediate
- Pathway: Others
- Pure form storage: -20°C for 3 years, 4°C for 2 years
- In solvent storage: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.