Learn More
Medchemexpress LLC 6-(3-fluorophenyl)-N-[1-(2,2,2-trifluoroethyl)piperidin-4-yl]pyridine-3-carboxamide | 1033836-12-2 | MFCD18206776 | 99.4% | 381.37 g/mol | C19H19F4N3O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1043910MG
Encompass_Preferred
HPGDS inhibitor 1 is a potent, selective small-molecule inhibitor of hematopoietic prostaglandin D synthase used in biochemical and cellular research to probe prostaglandin D2 pathways. It demonstrates sub-nanomolar enzyme potency and low-nanomolar cellular activity, with reported selectivity against related prostaglandin and eicosanoid enzymes.
  • Potent enzyme inhibition (IC50 ~0.6 nM).
  • Cellular activity around 32 nM in cell-based assays.
  • Selective versus human L-PGDS, mPGES, COX-1, COX-2, and 5-LOX.
  • High purity (99.4%).
  • Molecular formula C19H19F4N3O; molecular weight 381.37 g/mol.
  • Supplied in milligram pack sizes suitable for research use.
Catalog No. 50-002-41743