Learn More
Medchemexpress LLC 6-(3-(tert-butyl)-1H-pyrazol-5-yl)-N-(3-fluoro-4-methyl-2-(7-oxo-...)nicotinamide | 2729996-45-4 | 98.9% | 455.48 g/mol | C26H22FN5O2 | 5 MG

Supplier: Medchemexpress LLC HY1421605MG
GNE-9815 is a small-molecule Type II pan-RAF kinase inhibitor intended for preclinical research use only. It modulates RAF-mediated MAPK signaling and has been used in studies of KRAS-mutant cancers. The compound is supplied as a solid with high chemical purity and requires low-temperature storage.
- High purity (98.9%) suitable for biochemical and cell-based assays.
- Molecular weight 455.48 g/mol; formula C26H22FN5O2.
- Available in small research pack sizes, such as 5 mg.
- Storage recommendations: powder at -20°C; in solvent store at -80°C for long term.
- For research use only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.