Learn More
Medchemexpress LLC 6-[4-(1H-imidazol-1-yl)phenoxy]-N,N-dimethyl-1-hexanamine, dihydrochloride | 502656-68-0 | 98.0% | 10 MG

Supplier: Medchemexpress LLC HY120648A10MG
CAY10462 dihydrochloride is the dihydrochloride salt of CAY10434 and a potent, selective inhibitor of CYP4A (20-HETE synthase) used for research applications. Supplied as an off-white to light-yellow crystalline solid with high purity and reported solubility in DMSO.
- Potent inhibitor of CYP4A/20-HETE synthase.
- High purity (≈98%).
- Off-white to light-yellow crystalline solid appearance.
- Soluble in DMSO (reported 35.71 mg/mL with sonication/warming).
- Storage: -20°C for solid; in solvent -80°C for up to 6 months, -20°C for up to 1 month.
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.