Learn More
Medchemexpress LLC 6-(4-(5-(3,5-Difluorophenyl)-4,5-dihydro-1H-pyrazole-1-carbonyl)piperidin-1-yl)pyrimidine-4-carbonitrile | 99.9% | 396.39 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY114492A10MM/1ML
Encompass_Preferred
(Rac)-GSK547 is the racemate of GSK547, a highly selective and potent inhibitor of receptor-interacting serine/threonine protein kinase 1 (RIP1). It inhibits macrophage-mediated adaptive immune tolerance in pancreatic cancer.
  • Highly selective and potent inhibitor of receptor-interacting serine/threonine protein kinase 1 (RIP1)
  • Inhibits macrophage-mediated adaptive immune tolerance in pancreatic cancer
Catalog No. 50-004-37321