Learn More
Medchemexpress LLC 6-[6-(2-hexyldecanoyloxy)hexyl-(4-hydroxybutyl)amino]hexyl 2-hexyldecanoate | 2036272-55-4 | MFCD33402175 | 99.4% | C48H95NO5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13817010MG
Encompass_Preferred
ALC-0315 is an ionizable aminolipid used to formulate lipid nanoparticles (LNPs) for mRNA delivery research. It aids mRNA compaction, supports cellular uptake, and promotes endosomal destabilization to facilitate cytoplasmic release, making it useful for formulation and delivery studies.
  • Promotes mRNA compaction for efficient encapsulation.
  • Enhances cellular delivery and facilitates endosomal escape.
  • High analytical purity suitable for research applications.
  • Offered in small quantities appropriate for formulation development.
  • Compatible with standard lipid nanoparticle formulation workflows.
Catalog No. 50-002-60578