Learn More
Medchemexpress LLC 6,7-Dibromoquinoline-5,8-dione | 18633-05-1 | 95.5% | 316.94 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY24116
Encompass_Preferred
6,7-Dibromoquinoline-5,8-dione is a chemical compound. This solid compound has a molecular formula of C9H3Br2NO2 and a molecular weight of 316.94. It is typically stored as a powder at -20°C for 3 years or 4°C for 2 years, and in solvent at -80°C for 6 months or -20°C for 1 month.
  • Consistent with structure by 1H NMR and LCMS
  • Purity of 95.48% by LCMS
  • Available as a brown to reddish brown solid
  • Suitable for laboratory use and substance manufacturing
Catalog No. 50-004-02960