Learn More
Medchemexpress LLC 6,7-dihydro-8-(1-hydroxyethyl)-2-(pyridin-4-yl)-5H-pyrrolo[3,4-d]pyrimidin-4-amine, GP-82996, CINK4 | 359886-84-3 | 98.1% | 456.58 g/mol | C27H32N6O | 5 MG

Supplier: Medchemexpress LLC HY1367265MG
CGP-82996 is a small-molecule inhibitor of cyclin-dependent kinases CDK4 and CDK6 used in cellular and biochemical cancer research. It demonstrates inhibitory activity against CDK4/6 and can induce apoptosis in certain tumor cell lines (for example, U2OS).
- Targets CDK4 and CDK6 with reported IC50s of 1.5 μM (CDK4/cyclin D1) and 5.6 μM (CDK6/cyclin D1).
- Also shows activity against Cdk5/p35 (IC50 ≈ 25 μM).
- High purity (98.1%) suitable for research applications.
- Molecular formula C27H32N6O; molecular weight 456.58 g/mol.
- Soluble in DMSO and intended for in vitro research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.