Learn More
Medchemexpress LLC 6,7-dimethoxychromen-2-one | 120-08-1 | MFCD00006871 | 99.7% | 206.19 g/mol | C11H10O4 | 10 MG

Supplier: Medchemexpress LLC HYN022810MG
Scoparone (6,7-dimethoxychromen-2-one) is a naturally occurring coumarin isolated from Artemisia capillaris. Supplied as a high-purity solid for research use, it has reported anticoagulant, vasorelaxant, antioxidant, and anti-inflammatory activities and is commonly used in in vitro pharmacology and in vivo formulation studies.
- Purity 99.69% (as reported).
- Molecular weight 206.19 g/mol.
- Appearance solid, light yellow to yellow.
- Soluble in DMSO (50 mg/mL); in vivo formulation protocols available.
- Powder storage: -20°C for up to 3 years; in solvent store at -80°C for longer term.
- Intended for biochemical and pharmacological research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.