Learn More
Medchemexpress LLC 6,7-isoquinolinediol, 3,4-dihydro | 4602-83-9 | MFCD00143490 | 98.7% | 163.17 g/mol | C9H9NO2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW34154725MG
Encompass_Preferred
3,4-Dihydro-6,7-isoquinolinediol is a small-molecule β2-adrenergic receptor agonist intended for research use in studies of adrenergic signaling, inflammation, and immunology. Supplied as a high-purity solid, it is appropriate for in vitro pharmacology, biochemical assays, and screening applications.
  • High purity (≈98.7%) suitable for analytical and biological assays.
  • Characterized by molecular formula C9H9NO2 and molecular weight 163.17 g/mol.
  • Demonstrated β2-AR agonist activity useful for receptor pharmacology studies.
  • Available in small-scale quantities for screening and assay development.
  • Includes batch-specific certificate of analysis with purity and storage recommendations.
Catalog No. 50-002-80483