Learn More
Medchemexpress LLC 6-Aldehydoisoophiopogonone A | 112500-90-0 | 1 MG

Supplier: Medchemexpress LLC HYN79421MG
6-Aldehydoisoophiopogonone A is a homoisoflavonoid that can be isolated from Ophiopogon japonicus.
- Homoisoflavonoid compound
- Isolated from Ophiopogon japonicus
- Store at 4°C, protected from light
- When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.