Learn More
Medchemexpress LLC 6-amino-2-[(2S)-pentan-2-yl]oxy-9-(5-piperidin-1-ylpentyl)-7H-purin-8-one | 1207629-49-9 | 99.8% | 390.52 g/mol | C20H34N6O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11825010MG
Encompass_Preferred
GSK2245035 is a selective Toll-like receptor 7 (TLR7) agonist developed for intranasal research use. It preferentially stimulates type-I interferon responses and has been investigated in clinical studies of allergic rhinitis and allergen-induced asthmatic responses. Supplied as a solid research reagent for laboratory use.
  • Selective TLR7 agonist with preferential type-I interferon stimulation.
  • Investigated in clinical pharmacology for allergen and asthma models.
  • High purity: 99.79% reported by supplier.
  • Physical form: solid, off-white to light yellow.
  • Packaged for research: 10 MG single vial.
Catalog No. 50-002-26248