Learn More
Medchemexpress LLC 6-amino-4-(3-(hydroxymethyl)-5-(5-hydroxypent-1-yn-1-yl)phenyl)-4-isopropyl-3-methyl-1,4-dihydr... | 2204289-53-0 | 99.1% | C23H26N4O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13497810MG
Encompass_Preferred
(Rac)-SHIN2 is a racemic pyrazolopyran-derived small-molecule inhibitor of serine hydroxymethyltransferase (SHMT). It is used in biochemical and cellular studies to probe one-carbon metabolism and SHMT-related pathways, and has reported antibacterial activity with low cytotoxicity in Hep2 cells. Available as a solid and as solution formats for laboratory use.
  • Inhibits serine hydroxymethyltransferase (SHMT).
  • Racemic small-molecule pyrazolopyran derivative.
  • Demonstrated antibacterial activity with low Hep2 cytotoxicity.
  • Supplied as solid and as 10 mM solutions in DMSO.
  • Molecular formula C23H26N4O3; molecular weight 406.48 g/mol.
Catalog No. 50-002-60658