Learn More
Medchemexpress LLC 6-Aminocaproic acid-d6 | 1228656-08-3 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0236S1MG
Encompass_Preferred
6-Aminocaproic acid-d6 is a deuterium-labeled variant of 6-Aminocaproic acid. The unlabeled compound, 6-Aminocaproic acid (EACA), is a monoamino carboxylic acid that acts as a potent and orally active inhibitor of plasmin and plasminogen. It functions as a strong antifibrinolytic agent by competitively binding to lysine residues on plasminogen, thereby inhibiting plasmin formation and reducing fibrinolysis. This substance is utilized in research concerning bleeding disorders.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis via techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06927