Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-aminochrysene | 2642-98-0 | MFCD00003699 | 99.7% | 243.30 g/mol | C18H13N | 50 MG
SDP

Supplier:  Medchemexpress LLC HY10831550MG

Encompass_Preferred

6-Aminochrysene is an aromatic amine research compound (CAS 2642-98-0) used in laboratory studies. The light yellow solid has molecular formula C18H13N and a molecular weight of 243.30 g/mol. The supplier lists high purity, DMSO solubility data, storage recommendations for powder and solutions, and multiple small pack sizes for research use.

  • Light yellow to yellow solid appearance.
  • Molecular formula C18H13N and molecular weight 243.30 g/mol.
  • High purity (99.7%).
  • Soluble in DMSO (100 mg/mL); sonication recommended.
  • Recommended storage conditions provided for powder and in-solution forms.
  • Available in small research pack sizes, including 50 mg.

Catalog No. 50-002-43788


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.