Learn More
Medchemexpress LLC HY-W010342 100mg Medchemexpress, 6-Aminonicotinamide CAS:329-89-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW010342100MG
Encompass_Preferred
Medchemexpress, HY-W010342 100mg 6-Aminonicotinamide CAS:329-89-5 6-Aminonicotinamide, a potent antimetabolite of nicotinamide, is competitive NADP+-dependent enzyme glucose-6-phosphate dehydrogenase (G6PD) inhibitor (Ki=0.46 μM). 6-Aminonicotinamide interferes with glycolysis, resulting in ATP depletion and synergizes with DNA-crosslinking chemotherapy drugs, such as Cisplatin, in killing cancer cells[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3290