Learn More
Cayman Chemical 6-APBhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1107910 MG
Encompass_Preferred
An analytical reference standard categorized as a benzofuran; a metabolite of 6-MAPB; a potential metabolite 6-EAPB; has been found in seized samples suspected of containing NPS; intended for research and forensic applications
Catalog No. 50-295-1199