Learn More
Bachem 6-Azido-hexanoic acid
SDP
Supplier:  Bachem 4066756.0001
Encompass_Preferred
6-Azido-hexanoic acid, 79598-53-1, C6H11N3O2, Mr 157.17, 1g. e-Azidocaproic acid. Building block for Staudinger ligation as well as for "click chemistry". This highly selective reaction can be applied for the conjugation of peptides with various molecules and for the cyclization of peptides. The azido moiety may also be considered as a protected amino group which is deblocked under reductive conditions.
Catalog No. 50-260-669