Learn More
Medchemexpress LLC 6-azido-N-acetylgalactosamine-UDP | 1202854-87-2 | 100.0% | 632.37 | C17H26N6O16P2 | 10 MG

Supplier: Medchemexpress LLC HY15690510MG
6-Azido-N-acetylgalactosamine-UDP is a UDP-activated azido-modified sugar donor used as an enzymatic substrate in glycosylation assays, including beta-1,3-N-acetylhexosaminyltransferase (LgtA)-catalyzed glycosylation of lactose. The reagent is supplied as a white to off-white solid with high purity for biochemical research and storage at low temperatures.
- High purity (99.95%).
- Molecular weight 632.37 g/mol.
- Chemical formula C17H26N6O16P2.
- Physical form: solid, white to off-white.
- Suggested storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Available sizes include 5 mg-500 mg (larger sizes by quote).
- Intended use: enzymatic glycosylation studies and biochemical assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.