Learn More
Bellco Glass, Inc. 6 Baffle Shake Flask, 1000mL 38mm Delong Neck
SDP
Supplier:  Bellco Glass, Inc. 254361000
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. The 3 side and bottom baffles are used for optimal agitation. 38mm DeLong style neck.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 1LDimensions: 128x235mmClosure Sizes: 38Case Quantity: 6
Catalog No. 50-194-5538