Learn More
Medchemexpress LLC 6-Bromo-2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7(8H)-one | 1016636-76-2 | 99.7% | 342.62 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY32205
Encompass_Preferred
6-Bromo-2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7(8H)-one is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds.
  • Available as a solid, white to off-white appearance.
  • Can be stored at room temperature for up to 3 years.
  • When in solvent, store at -80°C for 2 years or -20°C for 1 year.
  • Intended for research purposes only.
Catalog No. 50-004-02885