Learn More
Medchemexpress LLC 6-Bromo-2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7(8H)-one | 1016636-76-2 | 99.7% | 342.62 | 50 G

Supplier: Medchemexpress LLC HY3220550G
6-Bromo-2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7(8H)-one is a laboratory chemical. It is intended for research use only and has not been fully validated for medical applications. This compound is used in the manufacture of substances and should be handled by qualified scientists in appropriately equipped facilities.
- High purity of 99.7%
- Molecular weight of 342.62
- Appears as a white to off-white solid
- Stable under recommended storage conditions
- Suitable for laboratory chemical applications
- Tested and complies with given specifications
- Store at room temperature for up to 3 years
- In solvent, store at -80°C for 2 years or -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.