Learn More
Medchemexpress LLC 6-Bromo-2-pyridinecarboxaldehyde | 34160-40-2 | 99.69% | C6H4BrNO | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY60194
Encompass_Preferred
6-Bromo-2-pyridinecarboxaldehyde is a biochemical reagent suitable for life science-related research, functioning as a biological material or organic compound. It has been utilized in studies involving rhodium-catalyzed reductive aldol coupling with divinyl ketones, which leads to syn-hydroxyenones. Additionally, it serves as a building block for Tris[(pyridyl)methyl]amine ligands.
  • Biochemical reagent for life science-related research.
  • Functions as a biological material or organic compound.
  • Utilized in studies involving rhodium-catalyzed reductive aldol coupling.
  • Serves as a building block for Tris[(pyridyl)methyl)amine ligands.
Catalog No. 50-003-58652